Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK057539.1 - 1 54 Chr. 21 - 32363021 - 32365281 [+] on Build GRCh38 85
GenBankmRNA DB030544.1 - 1 54 Chr. 21 - 32363021 - 32363570 [+] on Build GRCh38 85


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream melanocortin 2 receptor accessory protein MRAP B27, C21orf61, FALP, FGD2, GCCD2 -48.2kb Sense Chr. - 32291813 - 32314784 on Build GRCh38
Downstream small nucleolar RNA, H/ACA box 80A SNORA80A ACA67, SNORA80 11.9kb Anti-sense Chr. - 32377187 - 32377322 on Build GRCh38

View Full Product Details