The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145646.1 | NP_001139118.1 | 5 - 5 | 3131 | 93 |
NM_031301.3 | NP_112591.2 | 6 - 6 | 3254 | 93 | |
GenBank mRNA | AK057677.1 | - | 1 - 1 | 1652 | 93 |