Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD29
Entrez Gene ID
147463
Gene Name
ankyrin repeat domain 29
Gene Aliases
-
Chromosome Location
Chr.18: 23598926 - 23662914 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308238.1 NP_001295167.1 2 - 3 288 88
NM_173505.3 NP_775776.2 2 - 3 288 88
XM_011525830.2 XP_011524132.1 2 - 3 307 88
XM_017025561.1 XP_016881050.1 2 - 3 310 88
GenBank mRNA AK057782.1 - 2 - 3 218 88
AK313667.1 - 2 - 3 206 88
BC030622.1 - 2 - 3 293 88

View Full Product Details