Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD8
Entrez Gene ID
142891
Gene Name
sterile alpha motif domain containing 8
Gene Aliases
HEL-177, SMSr
Chromosome Location
Chr.10: 75099592 - 75182123 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174156.1 NP_001167627.1 3 - 4 780 80
NM_144660.2 NP_653261.1 3 - 4 836 80
XM_005269541.4 XP_005269598.1 3 - 4 971 80
XM_011539311.1 XP_011537613.1 3 - 4 899 80
XM_011539312.2 XP_011537614.1 3 - 4 829 80
XM_017015738.1 XP_016871227.1 3 - 4 836 80
XM_017015739.1 XP_016871228.1 3 - 4 864 80
GenBank mRNA AK057811.1 - 3 - 4 764 80
BC080593.1 - 3 - 4 780 80
EU668332.1 - 3 - 4 764 80
GQ472208.1 - 3 - 4 764 80

View Full Product Details