Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD16
Entrez Gene ID
118663
Gene Name
BTB domain containing 16
Gene Aliases
C10orf87
Chromosome Location
Chr.10: 122270745 - 122338180 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318189.1 NP_001305118.1 1 - 2 218 61
NM_144587.3 NP_653188.2 1 - 2 218 61
XM_011539239.2 XP_011537541.1 1 - 2 769 61
XM_011539240.2 XP_011537542.1 1 - 2 768 61
XM_011539241.2 XP_011537543.1 1 - 2 768 61
XM_011539242.2 XP_011537544.1 1 - 2 769 61
GenBank mRNA AK058088.1 - 1 - 2 208 61
BC109261.1 - 1 - 2 189 61
BC109262.1 - 1 - 2 189 61
DB052430.1 - 1 - 2 218 61

View Full Product Details