Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
-
Chromosome Location
Chr.2: 189674292 - 189763059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144708.3 NP_653309.3 20 - 21 3898 66
XM_011510673.2 XP_011508975.1 20 - 21 4417 66
XM_011510674.1 XP_011508976.1 20 - 21 4384 66
XM_011510675.2 XP_011508977.1 19 - 20 4312 66
XM_011510676.2 XP_011508978.1 20 - 21 4417 66
XM_011510677.2 XP_011508979.1 20 - 21 4417 66
XM_011510679.2 XP_011508981.1 18 - 19 3988 66
XM_011510685.2 XP_011508987.1 20 - 21 3973 66
XM_011510686.2 XP_011508988.1 16 - 17 3910 66
XM_011510687.2 XP_011508989.1 19 - 20 3781 66
XM_011510691.2 XP_011508993.1 16 - 17 2603 66
XM_011510692.2 XP_011508994.1 18 - 19 3617 66
XM_011510693.2 XP_011508995.1 17 - 18 3513 66
XM_011510694.2 XP_011508996.1 16 - 17 3332 66
XM_017003415.1 XP_016858904.1 15 - 16 3112 66
XM_017003416.1 XP_016858905.1 17 - 18 3524 66
XM_017003418.1 XP_016858907.1 15 - 16 3227 66
XM_017003419.1 XP_016858908.1 15 - 16 3194 66
XM_017003420.1 XP_016858909.1 15 - 16 3130 66
GenBank mRNA AJ549812.1 - 19 - 20 3597 66
AK058144.1 - 8 - 9 1250 66
BC044907.1 - 16 - 17 3391 66
BC047413.1 - 23 - 24 3462 66

View Full Product Details