Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBXN10
Entrez Gene ID
127733
Gene Name
UBX domain protein 10
Gene Aliases
UBXD3
Chromosome Location
Chr.1: 20183106 - 20196060 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152376.4 NP_689589.1 1 - 2 74 54
GenBank mRNA AK058158.1 - 1 - 2 59 54
BC047428.1 - 1 - 2 69 54
BX648631.1 - 1 - 2 64 54
DB061550.1 - 1 - 2 74 54

View Full Product Details