Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXorf58
Entrez Gene ID
254158
Gene Name
chromosome X open reading frame 58
Gene Aliases
-
Chromosome Location
Chr.X: 23907932 - 23939520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169574.1 NP_001163045.1 5 - 6 768 144
NM_152761.2 NP_689974.2 5 - 6 768 144
XM_011545473.2 XP_011543775.1 5 - 6 525 144
GenBank mRNA AK058173.1 - 5 - 6 768 144
BC117344.1 - 5 - 6 680 144
BC117346.1 - 5 - 6 680 144
BC144007.1 - 5 - 6 680 144

View Full Product Details