The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039577.3 | NP_001034666.1 | 5 - 6 | 692 | 95 |
NR_135154.1 | 5 - 6 | 692 | 95 | ||
XM_011542221.2 | XP_011540523.1 | 3 - 4 | 282 | 95 | |
XM_017002420.1 | XP_016857909.1 | 5 - 6 | 690 | 95 | |
XM_017002421.1 | XP_016857910.1 | 5 - 6 | 676 | 95 | |
XM_017002422.1 | XP_016857911.1 | 5 - 6 | 689 | 95 | |
GenBank mRNA | AF251294.1 | - | 3 - 4 | 328 | 95 |
AK057133.1 | - | 3 - 4 | 282 | 95 | |
AK074112.1 | - | 5 - 6 | 663 | 95 | |
AL834441.1 | - | 5 - 6 | 623 | 95 | |
BC016333.1 | - | 5 - 6 | 672 | 95 |