Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 40225469-40225983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRK2
Entrez Gene ID
120892
Gene Name
leucine rich repeat kinase 2
Gene Aliases
AURA17, DARDARIN, PARK8, RIPK7, ROCO2
UniGene
Hs.187636
Gene Chromosome Location
Chr.12: 40224895-40369285 on Build GRCh38
Transcripts
NM_198578.3 XM_005268629.3 XM_011537877.2 XM_011537881.2 XM_011537882.2 XM_017018786.1 XM_017018789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6233703 c.196A>G p.I66V Chr.12: 40225599-40225599 on Build GRCh38
6704374 c.183T>C p.H61H Chr.12: 40225586-40225586 on Build GRCh38
7275991 c.152-1G>T p.? Chr.12: 40225554-40225554 on Build GRCh38
7127096 c.199G>A p.V67I Chr.12: 40225602-40225602 on Build GRCh38
6328088 c.190C>T p.L64L Chr.12: 40225593-40225593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17465730 Chr.12: 40225910-40225910 on Build GRCh38
rs987708014 Chr.12: 40225778-40225778 on Build GRCh38
rs370488844 Chr.12: 40225572-40225572 on Build GRCh38
rs756020134 Chr.12: 40225932-40225932 on Build GRCh38
rs887954350 Chr.12: 40225620-40225620 on Build GRCh38
rs1041726930 Chr.12: 40225880-40225880 on Build GRCh38
rs376053849 Chr.12: 40225515-40225515 on Build GRCh38
rs762226610 Chr.12: 40225680-40225680 on Build GRCh38
rs545231732 Chr.12: 40225541-40225541 on Build GRCh38
rs894713683 Chr.12: 40225590-40225590 on Build GRCh38
rs201452811 Chr.12: 40225672-40225672 on Build GRCh38
rs756784783 Chr.12: 40225596-40225596 on Build GRCh38
rs997773703 Chr.12: 40225943-40225943 on Build GRCh38
rs557133869 Chr.12: 40225800-40225800 on Build GRCh38
rs199610083 Chr.12: 40225505-40225505 on Build GRCh38
rs546973780 Chr.12: 40225710-40225710 on Build GRCh38
rs745827398 Chr.12: 40225676-40225676 on Build GRCh38
rs150422099 Chr.12: 40225580-40225580 on Build GRCh38
rs2723273 Chr.12: 40225499-40225499 on Build GRCh38
rs967917787 Chr.12: 40225770-40225770 on Build GRCh38
rs748190697 Chr.12: 40225802-40225802 on Build GRCh38
rs202092317 Chr.12: 40225658-40225658 on Build GRCh38
rs545177721 Chr.12: 40225957-40225957 on Build GRCh38
rs763779128 Chr.12: 40225589-40225589 on Build GRCh38
rs1491944 Chr.12: 40225879-40225879 on Build GRCh38
rs772731036 Chr.12: 40225530-40225531 on Build GRCh38
rs200042985 Chr.12: 40225669-40225669 on Build GRCh38
rs776649677 Chr.12: 40225563-40225563 on Build GRCh38
rs72546335 Chr.12: 40225558-40225558 on Build GRCh38
rs17465737 Chr.12: 40225955-40225955 on Build GRCh38
rs910419502 Chr.12: 40225781-40225781 on Build GRCh38
rs762593111 Chr.12: 40225585-40225585 on Build GRCh38
rs1030323641 Chr.12: 40225733-40225733 on Build GRCh38
rs993145030 Chr.12: 40225939-40225939 on Build GRCh38
rs36223554 Chr.12: 40225872-40225872 on Build GRCh38
rs186083270 Chr.12: 40225684-40225684 on Build GRCh38
rs77259873 Chr.12: 40225504-40225504 on Build GRCh38
rs567679632 Chr.12: 40225668-40225668 on Build GRCh38
rs555075625 Chr.12: 40225894-40225894 on Build GRCh38
rs750946093 Chr.12: 40225618-40225618 on Build GRCh38
rs200809461 Chr.12: 40225665-40225665 on Build GRCh38
rs1022102499 Chr.12: 40225765-40225765 on Build GRCh38
rs1037928544 Chr.12: 40225925-40225925 on Build GRCh38
rs938804406 Chr.12: 40225893-40225893 on Build GRCh38
rs200860610 Chr.12: 40225677-40225677 on Build GRCh38
rs373126789 Chr.12: 40225511-40225511 on Build GRCh38
rs569380771 Chr.12: 40225854-40225854 on Build GRCh38
rs753309547 Chr.12: 40225766-40225767 on Build GRCh38
rs755218669 Chr.12: 40225664-40225664 on Build GRCh38
rs762584163 Chr.12: 40225581-40225598 on Build GRCh38
rs1041746016 Chr.12: 40225517-40225517 on Build GRCh38
rs202157354 Chr.12: 40225561-40225561 on Build GRCh38
rs903255008 Chr.12: 40225885-40225885 on Build GRCh38
rs17465723 Chr.12: 40225786-40225786 on Build GRCh38
rs750010972 Chr.12: 40225739-40225739 on Build GRCh38
rs370857512 Chr.12: 40225510-40225510 on Build GRCh38
rs762788995 Chr.12: 40225691-40225691 on Build GRCh38
rs971738090 Chr.12: 40225685-40225685 on Build GRCh38
rs869105616 Chr.12: 40225771-40225771 on Build GRCh38
rs192286003 Chr.12: 40225895-40225895 on Build GRCh38
rs367949688 Chr.12: 40225694-40225694 on Build GRCh38
rs988913748 Chr.12: 40225764-40225764 on Build GRCh38
rs942206250 Chr.12: 40225926-40225926 on Build GRCh38
rs201200707 Chr.12: 40225660-40225660 on Build GRCh38
rs777755566 Chr.12: 40225937-40225937 on Build GRCh38
rs200435213 Chr.12: 40225543-40225543 on Build GRCh38
rs201256581 Chr.12: 40225532-40225532 on Build GRCh38
rs754611183 Chr.12: 40225816-40225816 on Build GRCh38
rs75054132 Chr.12: 40225628-40225628 on Build GRCh38
rs1013522346 Chr.12: 40225924-40225924 on Build GRCh38
rs777012150 Chr.12: 40225519-40225519 on Build GRCh38
rs767872726 Chr.12: 40225616-40225616 on Build GRCh38
rs1002981390 Chr.12: 40225525-40225525 on Build GRCh38
rs2256286 Chr.12: 40225740-40225740 on Build GRCh38
rs776361465 Chr.12: 40225674-40225674 on Build GRCh38

View Full Product Details