Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72346405-72346900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEXA
Entrez Gene ID
3073
Gene Name
hexosaminidase subunit alpha
Gene Aliases
TSD
UniGene
Hs.604479
Gene Chromosome Location
Chr.15: 72343435-72376473 on Build GRCh38
Transcripts
NM_000520.5 NM_001318825.1 NR_134869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5659952 c.1171G>T p.V391L Chr.15: 72346686-72346686 on Build GRCh38
458974 c.1236C>T p.F412F Chr.15: 72346621-72346621 on Build GRCh38
6142824 c.1330G>A p.G444S Chr.15: 72346527-72346527 on Build GRCh38
701310 c.1182G>T p.E394D Chr.15: 72346675-72346675 on Build GRCh38
1239058 c.1273A>G p.I425V Chr.15: 72346584-72346584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777042785 Chr.15: 72346549-72346550 on Build GRCh38
rs868661831 Chr.15: 72346515-72346515 on Build GRCh38
rs140482769 Chr.15: 72346641-72346641 on Build GRCh38
rs373023700 Chr.15: 72346591-72346591 on Build GRCh38
rs1038092289 Chr.15: 72346625-72346625 on Build GRCh38
rs537494521 Chr.15: 72346505-72346505 on Build GRCh38
rs778878211 Chr.15: 72346668-72346668 on Build GRCh38
rs987672900 Chr.15: 72346462-72346462 on Build GRCh38
rs387906309 Chr.15: 72346579-72346580 on Build GRCh38
rs774562271 Chr.15: 72346593-72346593 on Build GRCh38
rs777737367 Chr.15: 72346675-72346675 on Build GRCh38
rs575836910 Chr.15: 72346684-72346684 on Build GRCh38
rs770303210 Chr.15: 72346674-72346674 on Build GRCh38
rs569827588 Chr.15: 72346501-72346501 on Build GRCh38
rs995463313 Chr.15: 72346451-72346451 on Build GRCh38
rs753737532 Chr.15: 72346761-72346761 on Build GRCh38
rs587779406 Chr.15: 72346552-72346552 on Build GRCh38
rs201471936 Chr.15: 72346604-72346604 on Build GRCh38
rs1020151571 Chr.15: 72346585-72346585 on Build GRCh38
rs267606862 Chr.15: 72346681-72346681 on Build GRCh38
rs760153692 Chr.15: 72346693-72346693 on Build GRCh38
rs752223090 Chr.15: 72346538-72346538 on Build GRCh38
rs775571839 Chr.15: 72346480-72346480 on Build GRCh38
rs371435283 Chr.15: 72346511-72346511 on Build GRCh38
rs760391947 Chr.15: 72346758-72346758 on Build GRCh38
rs780010367 Chr.15: 72346579-72346579 on Build GRCh38
rs768838512 Chr.15: 72346495-72346495 on Build GRCh38
rs774507567 Chr.15: 72346499-72346499 on Build GRCh38
rs765989303 Chr.15: 72346858-72346858 on Build GRCh38
rs761284736 Chr.15: 72346518-72346518 on Build GRCh38
rs1057516617 Chr.15: 72346707-72346707 on Build GRCh38
rs941638811 Chr.15: 72346482-72346482 on Build GRCh38
rs762494949 Chr.15: 72346620-72346620 on Build GRCh38
rs1800431 Chr.15: 72346551-72346551 on Build GRCh38
rs146714642 Chr.15: 72346691-72346691 on Build GRCh38
rs777212186 Chr.15: 72346692-72346692 on Build GRCh38
rs767041069 Chr.15: 72346526-72346526 on Build GRCh38
rs1003506073 Chr.15: 72346540-72346540 on Build GRCh38
rs369978649 Chr.15: 72346609-72346609 on Build GRCh38
rs773715120 Chr.15: 72346723-72346723 on Build GRCh38
rs121907958 Chr.15: 72346597-72346597 on Build GRCh38
rs121907963 Chr.15: 72346680-72346680 on Build GRCh38
rs1029249597 Chr.15: 72346457-72346457 on Build GRCh38
rs575121167 Chr.15: 72346626-72346626 on Build GRCh38
rs1800430 Chr.15: 72346662-72346662 on Build GRCh38
rs373783583 Chr.15: 72346714-72346714 on Build GRCh38
rs762663178 Chr.15: 72346756-72346756 on Build GRCh38
rs543414583 Chr.15: 72346748-72346748 on Build GRCh38
rs151251788 Chr.15: 72346628-72346628 on Build GRCh38
rs377513200 Chr.15: 72346759-72346759 on Build GRCh38
rs772879726 Chr.15: 72346689-72346689 on Build GRCh38
rs910063850 Chr.15: 72346697-72346697 on Build GRCh38
rs749368995 Chr.15: 72346582-72346582 on Build GRCh38
rs768164612 Chr.15: 72346703-72346703 on Build GRCh38
rs751189932 Chr.15: 72346468-72346468 on Build GRCh38
rs745772223 Chr.15: 72346567-72346567 on Build GRCh38
rs1057519465 Chr.15: 72346598-72346598 on Build GRCh38
rs1019027175 Chr.15: 72346471-72346471 on Build GRCh38
rs529223624 Chr.15: 72346852-72346853 on Build GRCh38
rs370266293 Chr.15: 72346679-72346679 on Build GRCh38
rs755973971 Chr.15: 72346661-72346661 on Build GRCh38
rs772431056 Chr.15: 72346601-72346601 on Build GRCh38
rs988192535 Chr.15: 72346689-72346689 on Build GRCh38
rs1035014194 Chr.15: 72346547-72346547 on Build GRCh38
rs535095877 Chr.15: 72346586-72346586 on Build GRCh38
rs766988109 Chr.15: 72346619-72346619 on Build GRCh38
rs756026917 Chr.15: 72346577-72346577 on Build GRCh38
rs752583618 Chr.15: 72346631-72346631 on Build GRCh38
rs145247513 Chr.15: 72346864-72346864 on Build GRCh38
rs941490300 Chr.15: 72346726-72346726 on Build GRCh38
rs564679855 Chr.15: 72346816-72346816 on Build GRCh38
rs540688425 Chr.15: 72346876-72346876 on Build GRCh38
rs374588958 Chr.15: 72346485-72346485 on Build GRCh38
rs757010519 Chr.15: 72346561-72346561 on Build GRCh38
rs953668823 Chr.15: 72346458-72346458 on Build GRCh38
rs751185874 Chr.15: 72346627-72346627 on Build GRCh38
rs758048549 Chr.15: 72346550-72346550 on Build GRCh38
rs755007226 Chr.15: 72346665-72346665 on Build GRCh38
rs202173526 Chr.15: 72346555-72346555 on Build GRCh38
rs761429006 Chr.15: 72346734-72346734 on Build GRCh38
rs968453006 Chr.15: 72346781-72346781 on Build GRCh38

View Full Product Details