Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7687294-7687802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TP53
Entrez Gene ID
7157
Gene Name
tumor protein p53
Gene Aliases
BCC7, LFS1, P53, TRP53
UniGene
Hs.437460
Gene Chromosome Location
Chr.17: 7668402-7687550 on Build GRCh38
Transcripts
NM_000546.5 NM_001126112.2 NM_001126113.2 NM_001126114.2 NM_001126118.1 NM_001276695.1 NM_001276696.1 NM_001276760.1 NM_001276761.1 NM_001143990.1 NM_001143991.1 XM_017024812.1

Gene Symbol
WRAP53
Entrez Gene ID
55135
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
UniGene
Hs.408312
Gene Chromosome Location
Chr.17: 7686071-7703502 on Build GRCh38
Transcripts
NM_000546.5 NM_001126112.2 NM_001126113.2 NM_001126114.2 NM_001126118.1 NM_001276695.1 NM_001276696.1 NM_001276760.1 NM_001276761.1 NM_001143990.1 NM_001143991.1 XM_017024812.1

SNP Detail

SNP ID Chromosome Location
rs1011636234 Chr.17: 7687417-7687417 on Build GRCh38
rs893294262 Chr.17: 7687379-7687379 on Build GRCh38
rs886053514 Chr.17: 7687421-7687421 on Build GRCh38
rs759020317 Chr.17: 7687618-7687618 on Build GRCh38
rs892070762 Chr.17: 7687509-7687509 on Build GRCh38
rs905894455 Chr.17: 7687437-7687437 on Build GRCh38
rs886053516 Chr.17: 7687450-7687450 on Build GRCh38
rs531575965 Chr.17: 7687396-7687396 on Build GRCh38
rs34285921 Chr.17: 7687680-7687680 on Build GRCh38
rs754358384 Chr.17: 7687674-7687674 on Build GRCh38
rs961276468 Chr.17: 7687446-7687446 on Build GRCh38
rs1035732864 Chr.17: 7687438-7687438 on Build GRCh38
rs574904138 Chr.17: 7687432-7687432 on Build GRCh38
rs879254146 Chr.17: 7687390-7687390 on Build GRCh38
rs1026796876 Chr.17: 7687468-7687468 on Build GRCh38
rs953705814 Chr.17: 7687678-7687678 on Build GRCh38
rs886053515 Chr.17: 7687442-7687442 on Build GRCh38
rs976382456 Chr.17: 7687672-7687672 on Build GRCh38
rs550104262 Chr.17: 7687696-7687696 on Build GRCh38
rs1040581725 Chr.17: 7687615-7687615 on Build GRCh38
rs1044886073 Chr.17: 7687419-7687419 on Build GRCh38
rs867621933 Chr.17: 7687410-7687410 on Build GRCh38
rs1010820110 Chr.17: 7687555-7687555 on Build GRCh38
rs587780065 Chr.17: 7687373-7687373 on Build GRCh38
rs1016840948 Chr.17: 7687776-7687776 on Build GRCh38
rs184299271 Chr.17: 7687656-7687656 on Build GRCh38
rs1054623097 Chr.17: 7687460-7687460 on Build GRCh38
rs751266961 Chr.17: 7687565-7687565 on Build GRCh38
rs17884410 Chr.17: 7687459-7687459 on Build GRCh38
rs985340142 Chr.17: 7687568-7687568 on Build GRCh38
rs542205900 Chr.17: 7687720-7687720 on Build GRCh38
rs1044752659 Chr.17: 7687428-7687428 on Build GRCh38
rs17885780 Chr.17: 7687664-7687664 on Build GRCh38
rs965141713 Chr.17: 7687669-7687669 on Build GRCh38
rs1005248192 Chr.17: 7687713-7687713 on Build GRCh38
rs1053179984 Chr.17: 7687368-7687368 on Build GRCh38

View Full Product Details