Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7688265-7688772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WRAP53
Entrez Gene ID
55135
Gene Name
WD repeat containing antisense to TP53
Gene Aliases
DKCB3, TCAB1, WDR79
UniGene
Hs.408312
Gene Chromosome Location
Chr.17: 7686071-7703502 on Build GRCh38
Transcripts
NM_001143990.1 NM_001143991.1 NM_001143992.1 NM_018081.2 XM_011523952.2 XM_017024812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
707852 c.64C>G p.P22A Chr.17: 7688712-7688712 on Build GRCh38
5460066 c.117A>G p.E39E Chr.17: 7688765-7688765 on Build GRCh38
5505405 c.73C>G p.P25A Chr.17: 7688721-7688721 on Build GRCh38
3970392 c.76T>C p.S26P Chr.17: 7688724-7688724 on Build GRCh38
6823334 c.40T>A p.C14S Chr.17: 7688688-7688688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560833428 Chr.17: 7688349-7688349 on Build GRCh38
rs755130675 Chr.17: 7688629-7688629 on Build GRCh38
rs867179605 Chr.17: 7688649-7688649 on Build GRCh38
rs549679591 Chr.17: 7688672-7688672 on Build GRCh38
rs548795605 Chr.17: 7688518-7688518 on Build GRCh38
rs896923407 Chr.17: 7688658-7688658 on Build GRCh38
rs761454111 Chr.17: 7688609-7688609 on Build GRCh38
rs764437508 Chr.17: 7688665-7688666 on Build GRCh38
rs772549847 Chr.17: 7688701-7688701 on Build GRCh38
rs986262623 Chr.17: 7688615-7688615 on Build GRCh38
rs17551150 Chr.17: 7688443-7688443 on Build GRCh38
rs564330662 Chr.17: 7688394-7688394 on Build GRCh38
rs140694361 Chr.17: 7688680-7688680 on Build GRCh38
rs564890743 Chr.17: 7688424-7688424 on Build GRCh38
rs960318586 Chr.17: 7688479-7688479 on Build GRCh38
rs759226877 Chr.17: 7688641-7688642 on Build GRCh38
rs896833793 Chr.17: 7688351-7688351 on Build GRCh38
rs750219808 Chr.17: 7688734-7688734 on Build GRCh38
rs888460319 Chr.17: 7688323-7688323 on Build GRCh38
rs750849403 Chr.17: 7688513-7688513 on Build GRCh38
rs967391051 Chr.17: 7688503-7688503 on Build GRCh38
rs1052258786 Chr.17: 7688402-7688402 on Build GRCh38
rs17883670 Chr.17: 7688404-7688404 on Build GRCh38
rs531543362 Chr.17: 7688357-7688357 on Build GRCh38
rs918768607 Chr.17: 7688519-7688519 on Build GRCh38
rs530208087 Chr.17: 7688516-7688516 on Build GRCh38
rs776077808 Chr.17: 7688729-7688729 on Build GRCh38
rs969094860 Chr.17: 7688467-7688467 on Build GRCh38
rs542836334 Chr.17: 7688294-7688294 on Build GRCh38
rs1052184865 Chr.17: 7688675-7688675 on Build GRCh38
rs923542780 Chr.17: 7688514-7688514 on Build GRCh38
rs750344241 Chr.17: 7688626-7688626 on Build GRCh38
rs1039607781 Chr.17: 7688336-7688336 on Build GRCh38
rs11539199 Chr.17: 7688550-7688550 on Build GRCh38
rs372820855 Chr.17: 7688601-7688601 on Build GRCh38
rs901125160 Chr.17: 7688341-7688341 on Build GRCh38
rs879058020 Chr.17: 7688557-7688557 on Build GRCh38
rs376286349 Chr.17: 7688663-7688663 on Build GRCh38
rs1057516027 Chr.17: 7688335-7688335 on Build GRCh38
rs951508354 Chr.17: 7688563-7688563 on Build GRCh38
rs942586705 Chr.17: 7688333-7688333 on Build GRCh38
rs990211123 Chr.17: 7688449-7688449 on Build GRCh38
rs761696793 Chr.17: 7688732-7688732 on Build GRCh38
rs932258006 Chr.17: 7688559-7688559 on Build GRCh38
rs930112339 Chr.17: 7688320-7688320 on Build GRCh38
rs748265946 Chr.17: 7688501-7688501 on Build GRCh38
rs779431250 Chr.17: 7688683-7688683 on Build GRCh38
rs1045842881 Chr.17: 7688669-7688669 on Build GRCh38
rs984243245 Chr.17: 7688593-7688593 on Build GRCh38
rs370181839 Chr.17: 7688410-7688410 on Build GRCh38
rs150282629 Chr.17: 7688694-7688694 on Build GRCh38
rs1043471019 Chr.17: 7688681-7688681 on Build GRCh38
rs752687945 Chr.17: 7688662-7688662 on Build GRCh38
rs775043934 Chr.17: 7688718-7688718 on Build GRCh38
rs768364856 Chr.17: 7688725-7688725 on Build GRCh38
rs886053517 Chr.17: 7688563-7688563 on Build GRCh38
rs765268037 Chr.17: 7688623-7688623 on Build GRCh38
rs754257593 Chr.17: 7688717-7688720 on Build GRCh38
rs760086957 Chr.17: 7688722-7688722 on Build GRCh38
rs1024209548 Chr.17: 7688376-7688376 on Build GRCh38
rs747300674 Chr.17: 7688311-7688311 on Build GRCh38
rs17880282 Chr.17: 7688679-7688679 on Build GRCh38
rs570354139 Chr.17: 7688642-7688642 on Build GRCh38
rs1048842870 Chr.17: 7688321-7688321 on Build GRCh38
rs79199718 Chr.17: 7688511-7688511 on Build GRCh38
rs781470763 Chr.17: 7688640-7688640 on Build GRCh38
rs547538493 Chr.17: 7688472-7688472 on Build GRCh38
rs376621917 Chr.17: 7688666-7688666 on Build GRCh38
rs765073967 Chr.17: 7688733-7688733 on Build GRCh38

View Full Product Details