Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8122939-8123449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HES7
Entrez Gene ID
84667
Gene Name
hes family bHLH transcription factor 7
Gene Aliases
SCDO4, bHLHb37
UniGene
Hs.434828
Gene Chromosome Location
Chr.17: 8120590-8126032 on Build GRCh38
Transcripts
NM_001165967.1 NM_032580.3 XM_011524041.2 XM_017025232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382540 c.100G>T p.E34* Chr.17: 8123069-8123069 on Build GRCh38
4070893 c.93C>G p.S31R Chr.17: 8123076-8123076 on Build GRCh38
5387713 c.96G>A p.L32L Chr.17: 8123073-8123073 on Build GRCh38
6322227 c.84C>G p.I28M Chr.17: 8123085-8123085 on Build GRCh38
3388326 c.70C>A p.R24S Chr.17: 8123099-8123099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765170412 Chr.17: 8123101-8123101 on Build GRCh38
rs1031718879 Chr.17: 8123284-8123284 on Build GRCh38
rs1051538192 Chr.17: 8123403-8123403 on Build GRCh38
rs139906985 Chr.17: 8122961-8122961 on Build GRCh38
rs371748968 Chr.17: 8122986-8122986 on Build GRCh38
rs534956126 Chr.17: 8123166-8123166 on Build GRCh38
rs761667617 Chr.17: 8123099-8123099 on Build GRCh38
rs886043538 Chr.17: 8123022-8123022 on Build GRCh38
rs568312773 Chr.17: 8123239-8123239 on Build GRCh38
rs760984393 Chr.17: 8123215-8123215 on Build GRCh38
rs938201039 Chr.17: 8123342-8123342 on Build GRCh38
rs143317942 Chr.17: 8123238-8123238 on Build GRCh38
rs775539610 Chr.17: 8123133-8123133 on Build GRCh38
rs759833400 Chr.17: 8123076-8123076 on Build GRCh38
rs928161530 Chr.17: 8123299-8123299 on Build GRCh38
rs956534860 Chr.17: 8123073-8123073 on Build GRCh38
rs1030608138 Chr.17: 8123210-8123210 on Build GRCh38
rs777996899 Chr.17: 8123140-8123140 on Build GRCh38
rs753408923 Chr.17: 8123094-8123094 on Build GRCh38
rs912479253 Chr.17: 8123138-8123138 on Build GRCh38
rs758157463 Chr.17: 8123117-8123117 on Build GRCh38
rs963939706 Chr.17: 8123294-8123294 on Build GRCh38
rs902919750 Chr.17: 8123398-8123398 on Build GRCh38
rs763576909 Chr.17: 8123091-8123091 on Build GRCh38
rs557357308 Chr.17: 8123258-8123258 on Build GRCh38
rs1003002087 Chr.17: 8123225-8123225 on Build GRCh38
rs986643107 Chr.17: 8123292-8123292 on Build GRCh38
rs930931905 Chr.17: 8123329-8123329 on Build GRCh38
rs915502099 Chr.17: 8123361-8123361 on Build GRCh38
rs1021612889 Chr.17: 8122975-8122975 on Build GRCh38
rs912414818 Chr.17: 8123293-8123293 on Build GRCh38
rs894721168 Chr.17: 8123231-8123231 on Build GRCh38
rs771978237 Chr.17: 8123126-8123127 on Build GRCh38
rs771634586 Chr.17: 8123058-8123058 on Build GRCh38
rs766234398 Chr.17: 8123130-8123130 on Build GRCh38
rs61731639 Chr.17: 8123046-8123046 on Build GRCh38
rs750219149 Chr.17: 8123038-8123038 on Build GRCh38
rs894626190 Chr.17: 8123400-8123400 on Build GRCh38
rs113994160 Chr.17: 8123096-8123096 on Build GRCh38
rs919852970 Chr.17: 8123321-8123321 on Build GRCh38
rs3027281 Chr.17: 8123259-8123259 on Build GRCh38
rs1021852812 Chr.17: 8123243-8123243 on Build GRCh38
rs890262203 Chr.17: 8123404-8123404 on Build GRCh38
rs948904509 Chr.17: 8123402-8123402 on Build GRCh38
rs773754529 Chr.17: 8123330-8123330 on Build GRCh38
rs774790059 Chr.17: 8123068-8123068 on Build GRCh38
rs1055275016 Chr.17: 8123357-8123357 on Build GRCh38
rs368725204 Chr.17: 8123013-8123013 on Build GRCh38
rs750181156 Chr.17: 8123110-8123110 on Build GRCh38
rs778047084 Chr.17: 8123050-8123050 on Build GRCh38
rs938461605 Chr.17: 8123387-8123387 on Build GRCh38
rs886865592 Chr.17: 8123224-8123224 on Build GRCh38
rs575713548 Chr.17: 8123269-8123269 on Build GRCh38
rs1057286113 Chr.17: 8123390-8123390 on Build GRCh38
rs376311641 Chr.17: 8123028-8123028 on Build GRCh38
rs372627919 Chr.17: 8123090-8123090 on Build GRCh38
rs772297813 Chr.17: 8123346-8123346 on Build GRCh38
rs919342333 Chr.17: 8123041-8123041 on Build GRCh38
rs538689772 Chr.17: 8123382-8123384 on Build GRCh38
rs752195493 Chr.17: 8122992-8122992 on Build GRCh38
rs930983491 Chr.17: 8123186-8123186 on Build GRCh38
rs552373254 Chr.17: 8123003-8123003 on Build GRCh38
rs954644488 Chr.17: 8122968-8122968 on Build GRCh38
rs535857171 Chr.17: 8123249-8123249 on Build GRCh38
rs975303757 Chr.17: 8123300-8123300 on Build GRCh38
rs781290618 Chr.17: 8123039-8123039 on Build GRCh38
rs1057492958 Chr.17: 8123228-8123228 on Build GRCh38
rs376704207 Chr.17: 8123285-8123285 on Build GRCh38
rs77130240 Chr.17: 8123183-8123183 on Build GRCh38
rs1039374525 Chr.17: 8123184-8123184 on Build GRCh38
rs756416284 Chr.17: 8123136-8123136 on Build GRCh38
rs376049573 Chr.17: 8123008-8123008 on Build GRCh38
rs987042061 Chr.17: 8123133-8123133 on Build GRCh38
rs374255567 Chr.17: 8123376-8123376 on Build GRCh38
rs1007405768 Chr.17: 8123405-8123405 on Build GRCh38
rs908281855 Chr.17: 8123377-8123377 on Build GRCh38
rs749800673 Chr.17: 8123055-8123055 on Build GRCh38
rs749382799 Chr.17: 8123163-8123163 on Build GRCh38
rs986116992 Chr.17: 8122971-8122971 on Build GRCh38
rs963338841 Chr.17: 8123263-8123263 on Build GRCh38
rs1011210823 Chr.17: 8123111-8123111 on Build GRCh38
rs370580036 Chr.17: 8123278-8123278 on Build GRCh38
rs368880766 Chr.17: 8123081-8123081 on Build GRCh38
rs371599866 Chr.17: 8123059-8123059 on Build GRCh38
rs919614925 Chr.17: 8123185-8123185 on Build GRCh38
rs573348922 Chr.17: 8123341-8123341 on Build GRCh38

View Full Product Details