The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001733.4 | NP_001724.3 | 10 - 11 | 1409 | 79 |
XM_017019915.1 | XP_016875404.1 | 10 - 11 | 1483 | 79 | |
GenBank mRNA | AK024951.1 | - | 8 - 9 | 2415 | 79 |
AK094009.1 | - | 3 - 4 | 1699 | 79 | |
AK222481.1 | - | 10 - 11 | 1403 | 79 | |
AK222491.1 | - | 10 - 11 | 1403 | 79 | |
AK291313.1 | - | 10 - 11 | 1431 | 79 | |
AK298445.1 | - | 10 - 11 | 1424 | 79 | |
AK303750.1 | - | 10 - 11 | 1302 | 79 | |
BC035220.1 | - | 10 - 11 | 1409 | 79 | |
M14058.1 | - | 10 - 11 | 1413 | 79 | |
X04701.1 | - | 10 - 11 | 1401 | 79 |