Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
Chromosome Location
Chr.19: 17824782 - 17848071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000215.3 NP_000206.2 17 - 18 2445 109
XM_005259896.3 XP_005259953.1 17 - 18 2484 109
GenBank mRNA U09607.1 - 16 - 17 2440 109
U31601.1 - 17 - 18 2440 109

View Full Product Details