Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWF
Entrez Gene ID
7450
Gene Name
von Willebrand factor
Gene Aliases
F8VWF, VWD
Chromosome Location
Chr.12: 5948874 - 6124675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000552.4 NP_000543.2 32 - 33 5875 63
GenBank mRNA AK297600.1 - 4 - 5 504 63
M10321.1 - 16 - 17 3404 63
X04385.1 - 32 - 33 5719 63

View Full Product Details