The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000552.4 | NP_000543.2 | 35 - 36 | 6317 | 64 |
GenBank mRNA | AK297600.1 | - | 7 - 8 | 946 | 64 |
M10321.1 | - | 19 - 20 | 3846 | 64 | |
X04385.1 | - | 35 - 36 | 6161 | 64 |