The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184.3 | NP_001175.2 | 13 - 14 | 2927 | 108 |
XM_011512924.1 | XP_011511226.1 | 13 - 14 | 2894 | 108 | |
XM_011512925.1 | XP_011511227.1 | 12 - 13 | 2702 | 108 | |
XM_011512926.2 | XP_011511228.1 | 13 - 14 | 2894 | 108 | |
XM_017006643.1 | XP_016862132.1 | 12 - 13 | 2702 | 108 | |
GenBank mRNA | U49844.1 | - | 13 - 14 | 2910 | 108 |
U76308.1 | - | 13 - 14 | 2884 | 108 | |
Y09077.1 | - | 13 - 14 | 2884 | 108 |