Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTNNB1
Entrez Gene ID
1499
Gene Name
catenin beta 1
Gene Aliases
CTNNB, MRD19, armadillo
Chromosome Location
Chr.3: 41199451 - 41240448 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098209.1 NP_001091679.1 14 - 15 2408 67
NM_001098210.1 NP_001091680.1 14 - 15 2408 67
NM_001904.3 NP_001895.1 14 - 15 2408 67
XM_005264886.2 XP_005264943.1 14 - 15 2422 67
XM_006712983.1 XP_006713046.1 14 - 15 2320 67
XM_006712984.1 XP_006713047.1 14 - 15 2320 67
XM_017005738.1 XP_016861227.1 14 - 15 2422 67
GenBank mRNA AB062292.1 - 9 - 10 1371 67
AB451264.1 - 12 - 13 2140 67
AB451392.1 - 12 - 13 2140 67
AK289932.1 - 15 - 16 2296 67
AK294778.1 - 15 - 16 2521 67
AK296790.1 - 14 - 15 2228 67
AK299954.1 - 14 - 15 2090 67
AK314776.1 - 14 - 15 2250 67
BC058926.1 - 14 - 15 2350 67
X87838.1 - 14 - 15 2354 67
Z19054.1 - 14 - 15 2340 67

View Full Product Details