Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPN
Entrez Gene ID
3249
Gene Name
hepsin
Gene Aliases
TMPRSS1
Chromosome Location
Chr.19: 35040506 - 35066573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002151.2 NP_002142.1 11 - 12 1153 119
NM_182983.2 NP_892028.1 10 - 11 1733 119
XM_005258838.4 XP_005258895.2 10 - 11 1231 119
XM_006723181.3 XP_006723244.2 10 - 11 1107 119
XM_017026731.1 XP_016882220.1 10 - 11 1406 119
XM_017026732.1 XP_016882221.1 9 - 10 968 119
GenBank mRNA AK095160.1 - 4 - 5 1329 119
AK293622.1 - 9 - 10 987 119
AK315655.1 - 10 - 11 1086 119
BC025716.1 - 10 - 11 1105 119
EU644753.1 - 9 - 10 908 119
M18930.1 - 11 - 12 1153 119
X07002.1 - 5 - 6 569 119
X07732.1 - 10 - 11 1733 119

View Full Product Details