Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPN
Entrez Gene ID
3249
Gene Name
hepsin
Gene Aliases
TMPRSS1
Chromosome Location
Chr.19: 35040506 - 35066573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002151.2 NP_002142.1 12 - 13 1294 64
NM_182983.2 NP_892028.1 11 - 12 1874 64
XM_005258838.4 XP_005258895.2 11 - 12 1372 64
XM_006723181.3 XP_006723244.2 11 - 12 1248 64
XM_017026731.1 XP_016882220.1 11 - 12 1547 64
XM_017026732.1 XP_016882221.1 10 - 11 1109 64
GenBank mRNA AK095160.1 - 5 - 6 1470 64
AK293622.1 - 10 - 11 1128 64
AK315655.1 - 11 - 12 1227 64
BC025716.1 - 11 - 12 1246 64
EU644753.1 - 10 - 11 1049 64
M18930.1 - 12 - 13 1294 64
X07002.1 - 6 - 7 710 64
X07732.1 - 11 - 12 1874 64

View Full Product Details