The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003254.2 | NP_003245.1 | 4 - 5 | 522 | 61 |
GenBank mRNA | A10416.1 | - | 4 - 5 | 392 | 61 |
AK074854.1 | - | 4 - 5 | 500 | 61 | |
AK311937.1 | - | 4 - 5 | 374 | 61 | |
BC000866.1 | - | 4 - 5 | 475 | 61 | |
BC007097.1 | - | 4 - 5 | 423 | 61 | |
BQ181804.1 | - | 2 - 3 | 61 | ||
BU857950.1 | - | - | 569 | 61 | |
CR407638.1 | - | 3 - 4 | 330 | 61 | |
CR541982.1 | - | 3 - 4 | 330 | 61 | |
M12670.1 | - | 4 - 5 | 378 | 61 | |
M59906.1 | - | 4 - 5 | 373 | 61 | |
S68252.1 | - | 3 - 4 | 333 | 61 | |
X02598.1 | - | 4 - 5 | 371 | 61 | |
X03124.1 | - | 4 - 5 | 392 | 61 |