The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201359.1 | NP_001188288.1 | 3 - 4 | 300 | 70 |
NM_005624.3 | NP_005615.2 | 3 - 4 | 300 | 70 | |
XM_011528177.2 | XP_011526479.1 | 4 - 5 | 501 | 70 | |
XM_017027119.1 | XP_016882608.1 | 4 - 5 | 467 | 70 | |
XM_017027120.1 | XP_016882609.1 | 4 - 5 | 396 | 70 | |
XM_017027121.1 | XP_016882610.1 | 4 - 5 | 342 | 70 | |
XM_017027122.1 | XP_016882611.1 | 4 - 5 | 479 | 70 | |
GenBank mRNA | AK292662.1 | - | 3 - 4 | 300 | 70 |
BC125136.1 | - | 2 - 3 | 193 | 70 | |
BC130561.1 | - | 2 - 3 | 235 | 70 | |
BC144463.1 | - | 2 - 3 | 235 | 70 | |
U86358.1 | - | 2 - 3 | 189 | 70 |