The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007051.2 | NP_008982.1 | 13 - 14 | 1716 | 78 |
XM_017000136.1 | XP_016855625.1 | 14 - 15 | 1433 | 78 | |
GenBank mRNA | AF094700.1 | - | 10 - 11 | 789 | 78 |
AF106798.1 | - | 13 - 14 | 1263 | 78 | |
AF132938.1 | - | 13 - 14 | 1312 | 78 | |
AF136173.1 | - | 13 - 14 | 1312 | 78 | |
AJ271408.1 | - | 13 - 14 | 1588 | 78 | |
AK094843.1 | - | 7 - 8 | 872 | 78 | |
AK292840.1 | - | 14 - 15 | 1326 | 78 | |
AK293659.1 | - | 6 - 7 | 687 | 78 | |
AL133631.1 | - | 10 - 11 | 976 | 78 | |
BC004970.1 | - | 13 - 14 | 1716 | 78 | |
BC067100.1 | - | 13 - 14 | 1540 | 78 |