The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000195.4 | NP_000186.2 | 14 - 15 | 1657 | 73 |
NM_001311345.1 | NP_001298274.1 | 13 - 14 | 1610 | 73 | |
NM_001322476.1 | NP_001309405.1 | 14 - 15 | 1586 | 73 | |
NM_001322477.1 | NP_001309406.1 | 14 - 15 | 1666 | 73 | |
NM_001322478.1 | NP_001309407.1 | 13 - 14 | 1487 | 73 | |
NM_001322479.1 | NP_001309408.1 | 13 - 14 | 1567 | 73 | |
NM_001322480.1 | NP_001309409.1 | 12 - 13 | 1325 | 73 | |
NM_001322481.1 | NP_001309410.1 | 12 - 13 | 1405 | 73 | |
NM_001322482.1 | NP_001309411.1 | 11 - 12 | 1306 | 73 | |
NM_001322483.1 | NP_001309412.1 | 13 - 14 | 1443 | 73 | |
NM_001322484.1 | NP_001309413.1 | 13 - 14 | 1523 | 73 | |
NM_001322485.1 | NP_001309414.1 | 12 - 13 | 1415 | 73 | |
NM_001322487.1 | NP_001309416.1 | 14 - 15 | 1709 | 73 | |
NM_001322489.1 | NP_001309418.1 | 12 - 13 | 1467 | 73 | |
XM_005269757.4 | XP_005269814.1 | 14 - 15 | 1599 | 73 | |
XM_017016170.1 | XP_016871659.1 | 11 - 12 | 1213 | 73 | |
XM_017016171.1 | XP_016871660.1 | 12 - 13 | 1420 | 73 | |
XM_017016172.1 | XP_016871661.1 | 11 - 12 | 1231 | 73 | |
GenBank mRNA | AL833734.1 | - | 3 - 4 | 309 | 73 |
U65676.1 | - | 14 - 15 | 1601 | 73 |