Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1C
Entrez Gene ID
775
Gene Name
calcium voltage-gated channel subunit alpha1 C
Gene Aliases
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
Chromosome Location
Chr.12: 1969677 - 2697949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000719.6 NP_000710.5 13 - 14 2203 59
NM_001129827.1 NP_001123299.1 13 - 14 2203 59
NM_001129829.1 NP_001123301.1 13 - 14 2203 59
NM_001129830.2 NP_001123302.2 13 - 14 2168 59
NM_001129831.1 NP_001123303.1 13 - 14 2203 59
NM_001129832.1 NP_001123304.1 13 - 14 2203 59
NM_001129833.1 NP_001123305.1 13 - 14 2203 59
NM_001129834.1 NP_001123306.1 13 - 14 2203 59
NM_001129835.1 NP_001123307.1 13 - 14 2203 59
NM_001129836.1 NP_001123308.1 13 - 14 2203 59
NM_001129837.1 NP_001123309.1 13 - 14 2203 59
NM_001129838.1 NP_001123310.1 13 - 14 2203 59
NM_001129839.1 NP_001123311.1 13 - 14 2203 59
NM_001129840.1 NP_001123312.1 13 - 14 2203 59
NM_001129841.1 NP_001123313.1 13 - 14 2203 59
NM_001129842.1 NP_001123314.1 13 - 14 2203 59
NM_001129843.1 NP_001123315.1 13 - 14 2203 59
NM_001129844.1 NP_001123316.1 13 - 14 2194 59
NM_001129846.1 NP_001123318.1 13 - 14 2203 59
NM_001167623.1 NP_001161095.1 13 - 14 2203 59
NM_001167624.2 NP_001161096.2 13 - 14 2168 59
NM_001167625.1 NP_001161097.1 13 - 14 2203 59
NM_199460.3 NP_955630.3 13 - 14 2168 59
XM_006719017.2 XP_006719080.1 13 - 14 2257 59
XM_011521020.2 XP_011519322.1 14 - 15 2332 59
XM_011521023.2 XP_011519325.1 14 - 15 2517 59
XM_017019926.1 XP_016875415.1 16 - 17 3639 59
XM_017019927.1 XP_016875416.1 16 - 17 3639 59
XM_017019928.1 XP_016875417.1 16 - 17 3639 59
XM_017019929.1 XP_016875418.1 16 - 17 3639 59
XM_017019930.1 XP_016875419.1 16 - 17 3639 59
XM_017019931.1 XP_016875420.1 16 - 17 3639 59
XM_017019932.1 XP_016875421.1 14 - 15 3444 59
XM_017019933.1 XP_016875422.1 14 - 15 3444 59
XM_017019934.1 XP_016875423.1 15 - 16 3519 59
XM_017019935.1 XP_016875424.1 15 - 16 3519 59
XM_017019936.1 XP_016875425.1 14 - 15 3444 59
XM_017019937.1 XP_016875426.1 14 - 15 3444 59
XM_017019938.1 XP_016875427.1 14 - 15 3444 59
XM_017019939.1 XP_016875428.1 14 - 15 3444 59
XM_017019940.1 XP_016875429.1 14 - 15 3444 59
XM_017019941.1 XP_016875430.1 14 - 15 3444 59
XM_017019942.1 XP_016875431.1 14 - 15 3444 59
XM_017019943.1 XP_016875432.1 14 - 15 3444 59
XM_017019944.1 XP_016875433.1 14 - 15 3444 59
XM_017019945.1 XP_016875434.1 14 - 15 3444 59
XM_017019946.1 XP_016875435.1 14 - 15 3444 59
XM_017019947.1 XP_016875436.1 14 - 15 3444 59
XM_017019948.1 XP_016875437.1 14 - 15 3444 59
XM_017019949.1 XP_016875438.1 14 - 15 3444 59
XM_017019950.1 XP_016875439.1 14 - 15 3444 59
XM_017019951.1 XP_016875440.1 14 - 15 3435 59
XM_017019952.1 XP_016875441.1 14 - 15 3444 59
XM_017019953.1 XP_016875442.1 13 - 14 2257 59
XM_017019954.1 XP_016875443.1 13 - 14 2257 59
XM_017019955.1 XP_016875444.1 15 - 16 3519 59
GenBank mRNA AB209016.1 - 11 - 12 1638 59
AF465484.1 - 13 - 14 2155 59
AJ224873.1 - 13 - 14 1890 59
AY830711.1 - 13 - 14 1890 59
AY830712.1 - 13 - 14 1890 59
AY830713.1 - 13 - 14 1890 59
BC146846.1 - 13 - 14 2009 59
L29529.1 - 12 - 13 1822 59
L29534.1 - 12 - 13 1816 59
L29536.1 - 12 - 13 1816 59
Z34809.1 - 13 - 14 1890 59
Z34810.1 - 13 - 14 1890 59
Z34811.1 - 13 - 14 1890 59
Z34812.1 - 13 - 14 1890 59
Z34813.1 - 13 - 14 1890 59
Z34814.1 - 13 - 14 1890 59
Z34815.1 - 13 - 14 1890 59
Z34816.1 - 13 - 14 1890 59
Z34817.1 - 13 - 14 1890 59
Z34818.1 - 13 - 14 1890 59
Z34819.1 - 13 - 14 1890 59
Z34820.1 - 13 - 14 1890 59
Z34821.1 - 13 - 14 1890 59
Z34822.1 - 13 - 14 1890 59
Z74996.1 - 13 - 14 1890 59

View Full Product Details