The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000844.3 | NP_000835.1 | 2 - 3 | 1006 | 57 |
NM_181874.2 | NP_870989.1 | 2 - 3 | 1006 | 57 | |
XM_017006272.1 | XP_016861761.1 | 2 - 3 | 472 | 57 | |
XM_017006273.1 | XP_016861762.1 | 2 - 3 | 472 | 57 | |
GenBank mRNA | AK312488.1 | - | 2 - 3 | 1006 | 57 |
BC136458.1 | - | 2 - 3 | 822 | 57 | |
BC136459.1 | - | 2 - 3 | 822 | 57 | |
X94552.1 | - | 2 - 3 | 970 | 57 |