The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002214.2 | NP_002205.1 | 4 - 5 | 1342 | 90 |
XM_011515393.2 | XP_011513695.1 | 5 - 6 | 1145 | 90 | |
XM_011515394.2 | XP_011513696.1 | 5 - 6 | 743 | 90 | |
XM_011515396.1 | XP_011513698.1 | 5 - 6 | 1414 | 90 | |
XM_017012178.1 | XP_016867667.1 | 5 - 6 | 1039 | 90 | |
XM_017012179.1 | XP_016867668.1 | 5 - 6 | 1498 | 90 | |
XM_017012180.1 | XP_016867669.1 | 5 - 6 | 1791 | 90 | |
XM_017012181.1 | XP_016867670.1 | 6 - 7 | 1610 | 90 | |
XM_017012182.1 | XP_016867671.1 | 5 - 6 | 725 | 90 | |
XM_017012183.1 | XP_016867672.1 | 5 - 6 | 682 | 90 | |
GenBank mRNA | AK295044.1 | - | 5 - 6 | 1414 | 90 |
BC002630.2 | - | 4 - 5 | 1342 | 90 | |
BT006739.1 | - | 4 - 5 | 637 | 90 | |
M73780.1 | - | 4 - 5 | 1317 | 90 |