Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD84
Entrez Gene ID
8832
Gene Name
CD84 molecule
Gene Aliases
LY9B, SLAMF5, hCD84, mCD84
Chromosome Location
Chr.1: 160541094 - 160579516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184879.1 NP_001171808.1 3 - 4 720 71
NM_001184881.1 NP_001171810.1 3 - 4 720 71
NM_001184882.1 NP_001171811.1 2 - 3 378 71
NM_003874.3 NP_003865.1 3 - 4 720 71
XM_011510094.2 XP_011508396.1 3 - 4 701 71
XM_011510095.2 XP_011508397.1 2 - 3 377 71
GenBank mRNA AF054815.1 - 3 - 4 710 71
AF054816.1 - 3 - 4 710 71
AF054817.1 - 3 - 4 710 71
AJ223324.1 - 3 - 4 685 71
AK296290.1 - 2 - 3 339 71
AK313496.1 - 3 - 4 686 71
BC020063.1 - 3 - 4 665 71
CR541847.1 - 3 - 4 641 71
CR933666.1 - 2 - 3 339 71
U82988.1 - 3 - 4 682 71
U96627.1 - 3 - 4 710 71
Y12632.1 - 3 - 4 710 71

View Full Product Details