Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBX2
Entrez Gene ID
6909
Gene Name
T-box 2
Gene Aliases
-
Chromosome Location
Chr.17: 61399896 - 61409466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005994.3 NP_005985.3 1 - 2 676 66
GenBank mRNA AB209378.1 - 1 - 2 207 66
AL832900.1 - 1 - 2 216 66
BC052566.1 - 1 - 2 676 66
U28049.1 - 1 - 2 412 66

View Full Product Details