Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CSF2RA
- Entrez Gene ID
-
1438
- Gene Name
- colony stimulating factor 2 receptor alpha subunit
- Gene Aliases
-
CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, SMDP4
- Chromosome Location
-
Chr.X: 1268800 - 1325097 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027760.1 |
|
9 - 10 |
1007 |
64 |
GenBank mRNA |
L29348.1 |
- |
9 - 10 |
962 |
64 |
View Full Product Details