The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000353.2 | NP_000344.1 | 4 - 5 | 505 | 135 |
GenBank mRNA | AK313380.1 | - | 4 - 5 | 506 | 135 |
BC130534.1 | - | 4 - 5 | 476 | 135 | |
X52520.1 | - | 4 - 5 | 502 | 135 | |
X55675.1 | - | 4 - 5 | 456 | 135 |