The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000445.4 | NP_000436.2 | 20 - 21 | 2436 | 107 |
NM_201378.3 | NP_958780.1 | 19 - 20 | 2321 | 107 | |
NM_201379.2 | NP_958781.1 | 19 - 20 | 2389 | 107 | |
NM_201380.3 | NP_958782.1 | 19 - 20 | 2887 | 107 | |
NM_201381.2 | NP_958783.1 | 19 - 20 | 2284 | 107 | |
NM_201382.3 | NP_958784.1 | 19 - 20 | 2374 | 107 | |
NM_201383.2 | NP_958785.1 | 19 - 20 | 2327 | 107 | |
NM_201384.2 | NP_958786.1 | 19 - 20 | 2435 | 107 | |
XM_005250976.3 | XP_005251033.1 | 20 - 21 | 2929 | 107 | |
XM_005250978.3 | XP_005251035.1 | 20 - 21 | 2454 | 107 | |
XM_005250979.4 | XP_005251036.1 | 20 - 21 | 2413 | 107 | |
XM_005250980.4 | XP_005251037.1 | 20 - 21 | 2498 | 107 | |
XM_005250981.3 | XP_005251038.1 | 20 - 21 | 2335 | 107 | |
XM_005250982.3 | XP_005251039.1 | 20 - 21 | 2328 | 107 | |
XM_005250983.2 | XP_005251040.1 | 19 - 20 | 2252 | 107 | |
XM_005250984.4 | XP_005251041.1 | 20 - 21 | 2317 | 107 | |
XM_006716588.3 | XP_006716651.1 | 21 - 22 | 2506 | 107 | |
XM_006716589.2 | XP_006716652.1 | 20 - 21 | 2267 | 107 | |
XM_006716590.3 | XP_006716653.1 | 20 - 21 | 2377 | 107 | |
XM_011517130.2 | XP_011515432.1 | 20 - 21 | 2442 | 107 | |
XM_011517131.2 | XP_011515433.1 | 19 - 20 | 2361 | 107 | |
XM_011517132.2 | XP_011515434.1 | 20 - 21 | 2450 | 107 | |
GenBank mRNA | AY480044.1 | - | 19 - 20 | 2393 | 107 |
AY480045.1 | - | 19 - 20 | 2264 | 107 | |
AY480046.1 | - | 19 - 20 | 2389 | 107 | |
AY480047.1 | - | 19 - 20 | 2887 | 107 | |
AY480048.1 | - | 19 - 20 | 2284 | 107 | |
AY480049.1 | - | 19 - 20 | 2313 | 107 | |
AY480050.1 | - | 19 - 20 | 2327 | 107 | |
AY480051.1 | - | 19 - 20 | 2435 | 107 | |
U53204.1 | - | 20 - 21 | 2438 | 107 |