Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOLR3
Entrez Gene ID
2352
Gene Name
folate receptor 3
Gene Aliases
FR-G, FR-gamma, gamma-hFR
Chromosome Location
Chr.11: 72135710 - 72139892 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000804.3 NP_000795.2 1 - 2 61 52
NM_001318045.1 NP_001304974.1 1 - 2 61 52
GenBank mRNA BC030285.1 - 1 - 2 61 52
BI836723.1 - 1 - 2 61 52

View Full Product Details