The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001972.3 | NP_001963.1 | 3 - 4 | 488 | 66 |
XM_011527775.1 | XP_011526077.1 | 4 - 5 | 1165 | 66 | |
XM_011527776.1 | XP_011526078.1 | 4 - 5 | 504 | 66 | |
GenBank mRNA | BC074816.2 | - | 3 - 4 | 405 | 66 |
BC074817.2 | - | 3 - 4 | 405 | 66 | |
D00187.1 | - | 2 - 3 | 278 | 66 | |
J03545.1 | - | 1 - 2 | 146 | 66 | |
M34379.1 | - | 3 - 4 | 406 | 66 | |
X05875.1 | - | 3 - 4 | 406 | 66 |