The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002715.2 | NP_002706.1 | 3 - 4 | 883 | 60 |
GenBank mRNA | AB451240.1 | - | 3 - 4 | 485 | 60 |
AB451365.1 | - | 3 - 4 | 485 | 60 | |
AK057424.1 | - | 3 - 4 | 615 | 60 | |
AK097599.1 | - | 3 - 4 | 695 | 60 | |
BC000400.2 | - | 3 - 4 | 684 | 60 | |
BC002657.2 | - | 3 - 4 | 684 | 60 | |
BC019275.1 | - | 3 - 4 | 684 | 60 | |
BC031696.1 | - | 3 - 4 | 684 | 60 | |
CR457417.1 | - | 3 - 4 | 485 | 60 | |
J03804.1 | - | 3 - 4 | 667 | 60 | |
X12646.1 | - | 3 - 4 | 568 | 60 |