The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003786.3 | NP_003777.2 | 29 - 30 | 4363 | 107 |
XM_005257763.3 | XP_005257820.1 | 28 - 29 | 4185 | 107 | |
XM_011525422.2 | XP_011523724.1 | 28 - 29 | 4290 | 107 | |
XM_011525423.1 | XP_011523725.1 | 29 - 30 | 4482 | 107 | |
XM_011525424.2 | XP_011523726.1 | 24 - 25 | 3713 | 107 | |
XM_011525425.2 | XP_011523727.1 | 28 - 29 | 4190 | 107 | |
XM_017025265.1 | XP_016880754.1 | 25 - 26 | 4226 | 107 | |
GenBank mRNA | AB010887.1 | - | 29 - 30 | 4297 | 107 |
AB208954.1 | - | 29 - 30 | 4303 | 107 | |
AF009670.1 | - | 29 - 30 | 4356 | 107 | |
AF083552.1 | - | 29 - 30 | 4300 | 107 | |
AF085690.1 | - | 29 - 30 | 4319 | 107 | |
AF085691.1 | - | 28 - 29 | 4468 | 107 | |
AF085692.1 | - | 28 - 29 | 4523 | 107 | |
AF104943.1 | - | 29 - 30 | 4287 | 107 | |
AF154001.1 | - | 4 - 5 | 1428 | 107 | |
BC137347.1 | - | 29 - 30 | 4314 | 107 | |
BC137348.1 | - | 29 - 30 | 4314 | 107 | |
U83659.1 | - | 7 - 8 | 1159 | 107 | |
Y17151.2 | - | 29 - 30 | 4325 | 107 |