The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005185.3 | NP_005176.1 | 1 - 1 | 547 | 100 |
GenBank mRNA | AK313934.1 | - | 1 - 1 | 475 | 100 |
BC031889.1 | - | 1 - 1 | 455 | 100 | |
M36707.1 | - | 1 - 1 | 1415 | 100 | |
M58026.1 | - | 1 - 1 | 475 | 100 |