The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005274.2 | NP_005265.1 | 1 - 2 | 148 | 91 |
GenBank mRNA | BM834854.1 | - | 1 - 2 | 148 | 91 |