The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006284.3 | NP_006275.1 | 3 - 4 | 510 | 64 |
GenBank mRNA | AL833496.1 | - | 1 - 2 | 565 | 64 |
BC012088.1 | - | 3 - 4 | 467 | 64 | |
BM990572.1 | - | 2 - 3 | 64 | ||
CR541943.1 | - | 3 - 4 | 455 | 64 | |
U13991.1 | - | 3 - 4 | 472 | 64 |