The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282716.1 | NP_001269645.1 | 25 - 26 | 2942 | 99 |
NM_001282717.1 | NP_001269646.1 | 25 - 26 | 3023 | 99 | |
NM_001282718.1 | NP_001269647.1 | 23 - 24 | 2816 | 99 | |
NM_012447.3 | NP_036579.2 | 25 - 26 | 3023 | 99 | |
XM_011515742.1 | XP_011514044.1 | 25 - 26 | 2990 | 99 | |
XM_017011683.1 | XP_016867172.1 | 25 - 26 | 2870 | 99 | |
XM_017011684.1 | XP_016867173.1 | 25 - 26 | 2990 | 99 | |
XM_017011685.1 | XP_016867174.1 | 25 - 26 | 3103 | 99 | |
XM_017011686.1 | XP_016867175.1 | 25 - 26 | 2870 | 99 | |
XM_017011687.1 | XP_016867176.1 | 23 - 24 | 2696 | 99 | |
GenBank mRNA | AJ007798.2 | - | 25 - 26 | 2943 | 99 |
AK292394.1 | - | 25 - 26 | 2850 | 99 | |
AK302587.1 | - | 22 - 23 | 2434 | 99 | |
AK302693.1 | - | 23 - 24 | 2639 | 99 | |
BC140932.1 | - | 25 - 26 | 2782 | 99 | |
BC146806.1 | - | 25 - 26 | 2848 | 99 |