Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM20B
Entrez Gene ID
9917
Gene Name
family with sequence similarity 20 member B
Gene Aliases
gxk1
Chromosome Location
Chr.1: 179025804 - 179076574 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324310.1 NP_001311239.1 4 - 5 759 70
NM_001324311.1 NP_001311240.1 4 - 5 769 70
NM_014864.3 NP_055679.1 4 - 5 864 70
XM_017003001.1 XP_016858490.1 5 - 6 810 70
XM_017003002.1 XP_016858491.1 5 - 6 803 70
GenBank mRNA AB007944.1 - 4 - 5 907 70
AB480690.1 - 4 - 5 864 70
AK289989.1 - 4 - 5 764 70
BC046441.1 - 4 - 5 732 70
BC051794.1 - 4 - 5 864 70

View Full Product Details