Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM10
Entrez Gene ID
56980
Gene Name
PR domain 10
Gene Aliases
PFM7
Chromosome Location
Chr.11: 129899706 - 130003097 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020228.2 NP_064613.2 15 - 16 2460 75
NM_199437.1 NP_955469.1 14 - 15 2448 75
NM_199438.1 NP_955470.1 11 - 12 2186 75
NM_199439.1 NP_955471.1 10 - 11 2174 75
XM_011542912.2 XP_011541214.1 14 - 15 2710 75
XM_017018023.1 XP_016873512.1 10 - 11 4259 75
GenBank mRNA AB033057.2 - 11 - 12 1970 75
AF275817.1 - 11 - 12 2186 75
AF503171.1 - 10 - 11 2105 75
AK000234.1 - 8 - 9 1415 75
AK310354.1 - 15 - 16 2320 75
AL833172.1 - 14 - 15 2451 75
BC112934.1 - 14 - 15 2440 75
BC117415.1 - 11 - 12 2025 75
BC143612.1 - 11 - 12 2025 75
BC143613.1 - 10 - 11 2013 75
BX648944.1 - 15 - 16 2461 75

View Full Product Details