Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNA7
Entrez Gene ID
3743
Gene Name
potassium voltage-gated channel subfamily A member 7
Gene Aliases
HAK6, KV1.7
Chromosome Location
Chr.19: 49067418 - 49072941 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031886.2 NP_114092.2 1 - 2 912 117
GenBank mRNA AF315818.1 - 1 - 2 593 117
AJ310479.1 - 1 - 2 912 117
AY779768.1 - 1 - 2 117
BC140906.1 - 1 - 2 117

View Full Product Details