The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077263.2 | NP_001070731.1 | 7 - 8 | 1095 | 68 |
NM_001206789.1 | NP_001193718.1 | 6 - 7 | 990 | 68 | |
NM_001206790.1 | NP_001193719.1 | 7 - 8 | 1095 | 68 | |
NM_001244995.1 | NP_001231924.1 | 7 - 8 | 1095 | 68 | |
GenBank mRNA | AB048796.1 | - | 7 - 8 | 1021 | 68 |
AB048797.1 | - | 6 - 7 | 946 | 68 | |
AK027798.1 | - | 7 - 8 | 1039 | 68 | |
AK300283.1 | - | 7 - 8 | 1007 | 68 | |
AY190317.1 | - | 7 - 8 | 1022 | 68 | |
BC018715.1 | - | 8 - 9 | 830 | 68 | |
BC114928.1 | - | 7 - 8 | 1014 | 68 |