The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000424.3 | NP_000415.2 | 4 - 5 | 1088 | 140 |
GenBank mRNA | AK093720.1 | - | 5 - 6 | 954 | 140 |
AK296823.1 | - | 4 - 5 | 401 | 140 | |
AK303972.1 | - | 5 - 6 | 918 | 140 | |
AK310827.1 | - | 4 - 5 | 1023 | 140 | |
BC024292.1 | - | 4 - 5 | 1012 | 140 | |
BC042132.1 | - | 4 - 5 | 1012 | 140 | |
BC071906.1 | - | 4 - 5 | 1007 | 140 | |
M19723.1 | - | 4 - 5 | 679 | 140 | |
M21389.1 | - | 4 - 5 | 1316 | 140 |