The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277115.1 | NP_001264044.1 | 14 - 15 | 2696 | 61 |
GenBank mRNA | AJ320497.1 | - | 14 - 15 | 2696 | 61 |