The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317227.1 | NP_001304156.1 | 2 - 3 | 693 | 57 |
NM_001317228.1 | NP_001304157.1 | 3 - 4 | 808 | 57 | |
NM_004061.4 | NP_004052.2 | 5 - 6 | 1322 | 57 | |
XM_011513927.2 | XP_011512229.1 | 3 - 4 | 599 | 57 | |
XM_017008920.1 | XP_016864409.1 | 4 - 5 | 949 | 57 | |
XM_017008921.1 | XP_016864410.1 | 5 - 6 | 1042 | 57 | |
GenBank mRNA | AK295123.1 | - | 3 - 4 | 808 | 57 |
AK314800.1 | - | 2 - 3 | 693 | 57 | |
BC047608.1 | - | 5 - 6 | 1027 | 57 | |
L33477.1 | - | 5 - 6 | 1323 | 57 | |
L34057.1 | - | 1 - 2 | 328 | 57 |