The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317227.1 | NP_001304156.1 | 7 - 8 | 1708 | 71 |
NM_001317228.1 | NP_001304157.1 | 7 - 8 | 1703 | 71 | |
NM_004061.4 | NP_004052.2 | 10 - 11 | 2337 | 71 | |
XM_011513927.2 | XP_011512229.1 | 8 - 9 | 1614 | 71 | |
XM_017008920.1 | XP_016864409.1 | 9 - 10 | 1964 | 71 | |
XM_017008921.1 | XP_016864410.1 | 10 - 11 | 2057 | 71 | |
GenBank mRNA | AK092176.1 | - | 5 - 6 | 987 | 71 |
AK295123.1 | - | 7 - 8 | 1703 | 71 | |
AK314800.1 | - | 7 - 8 | 1708 | 71 | |
BC047608.1 | - | 10 - 11 | 2042 | 71 | |
L33477.1 | - | 10 - 11 | 2338 | 71 |